Learn More
Euro Protection™ Gloves with Hydropellent™ Technology
Product Code. 12083016
Click to view available options
Size:
7
8
9
10
Experience high protection from contact cold with Euro Protection™ Gloves with Hydropellent™ Technology. The gloves are breathable, offer a good grip under wet/dry weather conditions, help handle slippery objects and are effective in cold and dark environments.
  • Latex
  • Disposable 
  • Fit to hand's shape
Specifications
Certifications/Compliance 3232, EN 420, EN 388, 020 EN 511, CE
Color Black
Material Latex
Size 10