Learn More
Thermo Scientific Chemicals [Glp1]-Apelin-13, human, bovine
For cellular and molecular biology applications
Brand:  Thermo Scientific Chemicals J66114.!D
 View more versions of this product
Product Code. 15427359
  • 71.40€ / 0.50mg
Estimated Shipment: 29-11-2024
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human, Bovine
-30°C to -10°C
1533.79
0.5 mg
Glp-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe
HPLC
Lyophilized Powder
[Glp1]-Apelin-13, human, bovine
RUO - research use only