missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GluR5/GRIK1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Specifications
Antigen | GluR5/GRIK1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18371127
|
Bio-Techne
NBP3-03819-20UL |
20 μL |
198.00€
20µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
18392146
|
Bio-Techne
NBP3-03819-100UL |
100 μL |
468.00€
100µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
Description
GluR5/GRIK1 Polyclonal specifically detects GluR5/GRIK1 in Mouse samples. It is validated for Western Blot.Specifications
GluR5/GRIK1 | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
EEA3, GLR5, GluR5, GluR-5, glutamate receptor, ionotropic, kainate 1, ionotropic kainate 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 859-918 of human GluR5/GRIK1 (NP_000821.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuronal Cell Markers, Neuroscience, Neurotransmission | |
PBS with 50% glycerol, pH7.3. | |
2897 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title