missing translation for 'onlineSavingsMsg'
Learn More
Glutaconic aldehyde dianilide hydrochloride, mixture of isomers, 98% Product Code.: 11478317

Glutaconic aldehyde dianilide hydrochloride, mixture of isomers, 98%

Product Code. 11478317
25 g, 25g
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11478317

Brand: Thermo Scientific Alfa Aesar A18390.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1497-49-0 | C17H17ClN2 | 284.79 g/mol

Glutaconic aldehyde dianilide hydrochloride is a tricarbocyanine laser dye intermediate. It is used as an intermediate for pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaconic aldehyde dianilide hydrochloride is a tricarbocyanine laser dye intermediate. It is used as an intermediate for pharmaceuticals and dyes.

Solubility
Soluble in methanol. Insoluble in water..

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1497-49-0
Molecular Formula C17H17ClN2
Molecular Weight (g/mol) 284.79
MDL Number MFCD00012630
InChI Key VUCMMJBDNXZQDJ-ZUJIUJENSA-N
Synonym hydrogen n-1e,3e-5-phenylimino penta-1,3-dien-1-yl aniline chloride
PubChem CID 5702667
IUPAC Name N-[(1E,3E)-5-phenyliminopenta-1,3-dienyl]aniline;hydrochloride
SMILES [H+].[Cl-].N(\C=C\C=C\C=NC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼124°C to 132°C
Quantity 25 g
Beilstein 3916924
Sensitivity Hygroscopic
Solubility Information Soluble in methanol. Insoluble in water..
Formula Weight 284.79
Percent Purity 98%
Chemical Name or Material Glutaconic aldehyde dianilide hydrochloride, mixture of isomers

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10236432 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog NumberA18390.06,A18390.14,A18390.22
Lot Number10228984 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog NumberA18390.06,A18390.14,A18390.22
Lot Number10236432 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18390.06,A18390.14,A18390.22
Lot Number10228984 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18390.06,A18390.14,A18390.22
Lot NumberG26R026 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18390.06,A18390.14,A18390.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glutaconic aldehyde dianilide hydrochloride, mixture of isomers, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.