missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutamine synthetase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
Antigen | Glutamine synthetase |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887833
|
Proteintech
11037-2-AP-20UL |
20 ÎĽL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877833
|
Proteintech
11037-2-AP-150UL |
150 ÎĽL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Structural maintece of chromosomes (SMC) and non-SMC condensin proteins associate into complexes that have been implicated in the process of chromosome condensation. A crucial prerequisite for accurate segregation of replicated sister chromatids is the condensation of the chromosomes into a manageable form prior to metaphase. The condensin I complex consists of two SMC subunits, SMC2 and SMC4, and three non-SMC subunits, CAP-H, CAP-G, and CAP-D2. An alternative complex, the condensin II complex, contains alternate non-SMC subunits, CAP-G2, CAP-H2, and CAP-D3. CAP-H2 is also known as Non-SMC condensin II complex, subunit H2 (NCAPH2) or kleisin beta isoform 2. The three non-SMC subunits in the condensing complexes form a regulatory subcomplex that is required to activate the SMC ATPases and to promote mitosis-specific chromatin binding of the holocomplex. The precise individual functions of each non-SMC protein in activation remain to be determined.Specifications
Glutamine synthetase | |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P09606, P15104, P15105 | |
14645, 24957, 2752 | |
Glutamine synthetase Fusion Protein Ag1510 | |
Primary | |
-20°C | |
GLUL |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GLNS, GLUL, Glutamate ammonia ligase, Glutamate decarboxylase, Glutamine synthetase, GS, PIG43, PIG59 | |
GLUL | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Glutamine synthetase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech