missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Reductase Rabbit anti-Human, Clone: S08-2E4, Novus Biologicals™
Rabbit Monoclonal Antibody
290.00€ - 588.00€
Specifications
Antigen | Glutathione Reductase |
---|---|
Clone | S08-2E4 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18392325
|
Bio-Techne
NBP3-15033-25UL |
25 μg |
290.00€
25µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
18396986
|
Bio-Techne
NBP3-15033-100UL |
100 μg |
588.00€
100µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
Description
Glutathione Reductase Monoclonal antibody specifically detects Glutathione Reductase in Human samples. It is validated for Western BlotSpecifications
Glutathione Reductase | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
2936 | |
IgG | |
Affinity-purified |
S08-2E4 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 1.8.1, EC 1.8.1.7, GLUR, glutathione reductase, glutathione reductase, mitochondrial, GR, GRase, GRD1, MGC78522 | |
A synthetic peptide of human Glutathione Reductase (Uniprot#: P00390) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title