Learn More
Glycine tert-butyl ester hydrochloride, 99%, Thermo Scientific Chemicals
27.14€ - 102.51€
Chemical Identifiers
CAS | 27532-96-3 |
---|---|
Molecular Formula | C6H14NO2 |
Molecular Weight (g/mol) | 132.18 |
MDL Number | MFCD00058255 |
InChI Key | SJMDMGHPMLKLHQ-UHFFFAOYSA-O |
Synonym | h-gly-otbu.hcl, tert-butyl 2-aminoacetate hydrochloride, glycine tert-butyl ester hydrochloride, glycine tert butyl ester hydrochloride, glycine tert-butyl ester hcl, glycine t-butyl ester hydrochloride, tert-butyl glycine hydrochloride, t-butyl glycinate, tert-butyl aminoacetate hydrochloride, glycine tert-butylester hydrochloride |
PubChem CID | 2725036 |
IUPAC Name | tert-butyl 2-aminoacetate;hydrochloride |
SMILES | CC(C)(C)OC(=O)C[NH3+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11353956
|
Thermo Scientific Alfa Aesar
L17116.03 |
1 g |
27.14€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363956
|
Thermo Scientific Alfa Aesar
L17116.06 |
5 g |
102.51€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.
Solubility
Soluble in methanol and water
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
27532-96-3 | |
132.18 | |
SJMDMGHPMLKLHQ-UHFFFAOYSA-O | |
2725036 | |
CC(C)(C)OC(=O)C[NH3+] |
C6H14NO2 | |
MFCD00058255 | |
h-gly-otbu.hcl, tert-butyl 2-aminoacetate hydrochloride, glycine tert-butyl ester hydrochloride, glycine tert butyl ester hydrochloride, glycine tert-butyl ester hcl, glycine t-butyl ester hydrochloride, tert-butyl glycine hydrochloride, t-butyl glycinate, tert-butyl aminoacetate hydrochloride, glycine tert-butylester hydrochloride | |
tert-butyl 2-aminoacetate;hydrochloride |
Specifications
∽140°C (decomposition) | |
C6H14NO2 | |
3989048 | |
Soluble in methanol and water | |
CC(C)(C)OC(=O)C[NH3+] | |
132.18 | |
167.64 | |
99% |
27532-96-3 | |
MFCD00058255 | |
h-gly-otbu.hcl, tert-butyl 2-aminoacetate hydrochloride, glycine tert-butyl ester hydrochloride, glycine tert butyl ester hydrochloride, glycine tert-butyl ester hcl, glycine t-butyl ester hydrochloride, tert-butyl glycine hydrochloride, t-butyl glycinate, tert-butyl aminoacetate hydrochloride, glycine tert-butylester hydrochloride | |
SJMDMGHPMLKLHQ-UHFFFAOYSA-O | |
tert-butyl 2-aminoacetate;hydrochloride | |
2725036 | |
Hygroscopic | |
Glycine tert-butyl ester hydrochloride |
Safety and Handling
missing translation for 'einecsNumber' : 214-223-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.