missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMO Extraction Kit
GMO Extraction Kit
Brand: Thermo Scientific Oxoid 4466336
919.79 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
The GMO Extraction Kit enables fast and easy purification of DNA for use in food safety testing for genetically modified organisms. The kit is capable of isolating highly purified DNA from a wide variety of different sample types without the use of toxic reagents. The benefits of the GMO Extraction Kit over other commercially-available products include:The GMO Extraction Kit employs silica-based DNA purification using spin column technology to provide samples for GMO testing. The kit has been specifically adapted for transgenic material analysis of a wide array of sample matrices, including raw materials, processed food, and feed. Effectively removing inhibitors, the GMO Extraction Kit provides isolated, highly-purified DNA, quickly, reliably, and safely.
The development and manufacture of this kit occurs through a partnership with IMEGEN (Instituto de Medicina Genomica), which has a long history of GMO research and development.
Other related products include:
Taqman™ Roundup Ready Soya Quantification Kit
TaqMan™ GMO Screening Kit
TaqMan™ GMO Maize Quantification Kit

Specifications
GMO Extraction Kit, 50 preps, PCR, Real-Time Quantitative PCR (qPCR) Sequencing Downstream Application, Enables Fast and Easy Purification of DNA For Use in Food Safety Testing For Genetically Modified Organisms | |
Food and Environmental, Fungi, Plant, Yeast | |
PCR, real-time quantitative PCR (qPCR), sequencing | |
50 Isolations | |
5 to 20g |
Silica Spin Column | |
Genomic DNA | |
Lysis buffer-1 2x500mL, lysis buffer-2 30mL, wash buffer-1 30mL, wash buffer-2 35mL, water 30mL, proteinase buffer 1.8mL, proteinase K 30mg, RNase 10mg, DNA filter columns 50, collection tubes 100 | |
GMO Extraction Kit | |
100ng/μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GMO Extraction Kit > 50 preps
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction