missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human Kappa Light Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69229
This item is not returnable.
View return policy
Description
Kappa Light Chain Polyclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoblot, Dot BlotSpecifications
Kappa Light Chain | |
Polyclonal | |
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoblotting, Dot Blot | |
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
A pool of purified Bence Jones kappa proteins isolated from human urine. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
10 mg | |
Adaptive Immunity, Immunology | |
3514 | |
Human | |
Lyophilized |
Western Blot, ELISA, Immunofluorescence, Immunoblot, Dot Blot | |
Unconjugated | |
Lyophilized from PBS (pH 7.2) | |
Goat | |
Ion exchange chromatography | |
RUO | |
Secondary | |
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |