missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human Lambda Light Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69775
This item is not returnable.
View return policy
Description
Lambda Light Chain Polyclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Western Blot, ELISA, Dot BlotSpecifications
Lambda Light Chain | |
Polyclonal | |
Lyophilized from PBS (pH 7.2) | |
Goat | |
Ion exchange chromatography | |
RUO | |
Secondary | |
Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Dot Blot | |
Unconjugated | |
IGLC, immunoglobulin lambda constant group | |
A pool of purified Bence Jones lambda proteins isolated from human urine. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
10 mg | |
Adaptive Immunity, Immunology | |
3537 | |
Human | |
Lyophilized |