missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Mouse IgG2b Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP2-68515

Product Code. 18617456

  • 421.00€ / 1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IgG2b Polyclonal antibody specifically detects IgG2b in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG2b
Polyclonal
Western Blot, ELISA 1:4000 - 1:8000, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Goat
Affinity purified
RUO
Secondary
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
HRP
PBS, 50% Glycerol
Mouse IgG2b paraproteins
1 mL
Neuronal Cell Markers, Neuroscience, Neurotransmission
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Goat anti-Mouse IgG2b Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™ > 1 mL; HRP

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.