missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Porcine IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72585
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
Specifications
IgG (H+L) | |
Polyclonal | |
Western Blot 1:500 - 1:2500, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:200 - 1:1000, Dot Blot | |
Goat | |
Multi-step | |
RUO | |
Secondary | |
Store at 4°C. Do not freeze. |
Western Blot, ELISA, Immunohistochemistry, Dot Blot | |
Alkaline Phosphatase (AP) | |
0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
Goat anti-Porcine IgG (H+L) Secondary Antibody [Alkaline Phosphatase] was produced by repeated immunization with Swine IgG whole molecule in goat. | |
1 mg | |
Adaptive Immunity, Immunology | |
Pig | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Goat anti-Porcine IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™ > 1 mg; Alkaline Phosphatase
Spot an opportunity for improvement?Share a Content Correction