missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Rat IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 647, Invitrogen™

Goat Polyclonal Secondary Antibody

373.00€

Specifications

Antigen Rat IgG (H+L) Highly Cross-Adsorbed
Concentration 2 mg/mL
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor Plus 647
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17129078
View Documents
Invitrogen
A48265
1 mg
373.00€
1mg
In Stock
to see stock.
Add to basket
 
Description

Description

To minimize cross-reactivity, the goat anti-rat IgG whole antibodies have been cross-adsorbed against serum proteins from mouse, horse, rabbit, bovine, and human to increase specificity of the antibody resulting in less background staining and cross-reactivity. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only nonspecific-binding secondary antibodies are captured in the column, and highly specific secondaries flow through. Further passages through additional columns result in highly cross-adsorbed preparations of secondary antibody. The benefits of these extra steps are apparent in multiplexing/multicolor-staining experiments where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experimentto eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically. This antibody binds to heavy chains on rat IgG and light chains on all rat immunoglobulins, does not bind non-immunoglobulin rat serum proteins or serum proteins/IgG from mouse, horse, rabbit, bovine, or human.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Rat IgG (H+L) Highly Cross-Adsorbed
Immunocytochemistry, Western Blot
Alexa Fluor Plus 647
Goat
Rat
Gamma Immunoglobins Heavy and Light chains
Secondary
4° C, store in dark
2 mg/mL
Polyclonal
Liquid
RUO
proprietary buffer with 0.016% Bromonitrodioxane, 0.016% Methylisothiazolone; pH 6.5
IgG
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications