missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG1 Heavy Chain Secondary Antibody, DyLight 755, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7117IR
This item is not returnable.
View return policy
Description
IgG1 Heavy Chain Polyclonal antibody specifically detects IgG1 Heavy Chain in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
IgG1 Heavy Chain | |
Polyclonal | |
50mM Sodium Borate | |
Rat IgG1 | |
1 mL | |
Secondary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
Goat | |
Immunogen affinity purified | |
RUO | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Goat anti-Rat IgG1 Heavy Chain Secondary Antibody, DyLight 755, Novus Biologicals™ > 1 mL; DyLight 755
Spot an opportunity for improvement?Share a Content Correction