Learn More
Gosselin™ 100x15mm Petri Dish with three vents, Optimized for automation
Optimized for automation.
Brand: Gosselin™ SB93-102
61.27 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Gosselin 100 x 15 mm Petri dishes have been designed in collaboration with Petri dish filling equipment manufacturers . These Petri dishes are ideal for microbiological examination either by surface plating or plate pouring techniques.- Crystal polystyrene
- Three vents
- Optimal flatness
- Manufactured in class 100
- SAL 10-6
- Easy-opening sleeves
Specifications
Polystyrene | |
Sterile | |
3 | |
90 mm |
Round | |
Optimized for automation | |
14.2 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
Product Content Correction
Product Title
Gosselin™ 100x15mm Petri Dish with three vents, Optimized for automation > Sterile