Learn More
Gosselin™ Natural LDPE Seals
Brand:  Gosselin OB65-01
Product Code. 11797145
  • 219.48€ / Pack of 1500
Only null left
Estimated Shipment: 05-09-2025
Specifications
Natural
1500/Carton
58 mm
LDPE
EC N°1935/2004
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Gosselin™ Natural LDPE Seals > O.D.: 58mm