Learn More
Gosselin™ Square HDPE Bottle, 2 L, Graduated, Sterile
Product Code. 16525104
Click to view available options
2 L; Sterile; Graduated; Square: HDPE
Applications: Environmental and Water Testing, Food and Beverage Testing, Pharmaceutical Quality Control and Life Sciences
  • Break-resistant and translucent high-density polyethylene (HDPE)
  • HDPE cap
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2 L
Color Clear
Description Assembled
Material HDPE
Quantity 50/Cs.
Sterility Sterile
Closure Color White
Graduated Yes
Shape Square
Closure Size 58 mm
Closure Type Cap with Seal