Learn More
Gosselin™ Straight Red Screw Cap Container, PP, Natural, Graduated, Sterile
Click to view available options
125mL
Description
Gosselin™ straight containers are designed for sample storage and transportation to laboratories.
- Clear polypropylene
- PE screw cap
Specifications
Specifications
Capacity (Metric) | 125 mL |
Capacity (English) | 4.2 oz. |
Color | Natural |
Sterility | Sterile |
Closure Color | Red |
Graduated | Yes |
Shape | Straight-sided, Round |
Closure Material | Polypropylene |
Closure Type | Screw Cap |