Learn More
Gosselin™ Gosselin™ Winged Plug
Suitable for different usage of tubes and applications
Brand: Gosselin™ BA110-20
177.54 EUR valid until 2025-03-14
Use promo code "24094" to get your promotional price.
Description
- Manufacturing proccess: All closures and tubes are produced with high-tech equipment using injection molding technology. The raw plastics used are virgin and comply with the current environmental standards. Each production lot is subject to inspection throughout the entire production process according to the standard references, which are reinforced by Gosselin’s own specifications.
- Tubes shapes: Wide range of tubes: round-bottom, conical-bottom, flat-bottom, winged bottom, can be used with the different analysers is on the market.
- Resistance: Most of our tubes are designed to withstand centrifugation up to a maximum at 3000 g with a suitable adaptor in a balanced rotor.
- Closures: Our closures are suitable for different usage of tubes and applications. That’s why we offer straight, winged, or screw caps. All closures are vertically ribbed for easier grip.
- Packaging: All tubes are protected with a polyethylene outer bag placed inside the boxes beforehand to prevent any risk of particle contamination.
- Product traceability: Thanks to the product label on each box, we can trace each production cycle. This label includes lot number and expiry date if the product has been sterilized.
- Raw materials: Compliant with the European directives in force, especially the absence of heavy metals (CE N°1935/2004), food compatibility (94/62/EC) and CE marking relating to medical devices for in vitro diagnostics (98/79/EC).
Specifications
LDPE | |
15000/Cs. | |
2500/Pk. |
Special Offers
-
Multiple Manufacturers
Product Content Correction
Product Title
Gosselin™ Gosselin™ Winged Plug > Pink, LDPE, 2500/Bag, 15000/Case