Learn More
Sampling Systems™ Graduated SteriWare Beakers
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
2 L
Sterility:
Non-sterile
Sterile
Description
- Made from food grade, FDA and EC compliant polypropylene
- Available sterile and non-sterile
- Wide range of sizes
- 100% manufactured and packed in a cleanroom
Specifications
Specifications
Capacity (Metric) | 25 mL |
Material | Polypropylene |
Sterility | Non-sterile |
Autoclavable | Autoclavable |
Disposable | Single-use |
Graduated | Yes |
Certifications/Compliance | FDA |
Scale | Molded Scale |
Style | Low Form |
Product Content Correction
Product Title
Sampling Systems™ Graduated SteriWare Beakers >