Learn More
Deutsch Neumann™ Grey Natural Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Grey Natural Rubber Stoppers are suitable for general use.
Brand:  Deutsch Neumann™ 1010164
 View more versions of this product
Product Code. 10140511
  • 7.95€ / Each
Only null left
Estimated Shipment: 10-07-2025
Description
  • Acc. to DIN 12 871
  • Durometer 40 Shore A
  • Available in a wide range of sizes
Specifications
Specifications
55mm
Gray
75.5 mm
Tapered Stopper
1/Ea.
64.5 mm
Natural Rubber
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Deutsch Neumann™ Grey Natural Rubber Stoppers > Bottom Diameter: 64.5mm; Height: 55mm