Learn More
Deutsch Neumann™ Grey Natural Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Grey Natural Rubber Stoppers are suitable for general use.
Brand:  Deutsch Neumann™ 1010150
 View more versions of this product
Product Code. 10577941
  • 4.55€ / Each
Only null left
Estimated Shipment: 20-06-2025
Description
  • Acc. to DIN 12 871
  • Durometer 40 Shore A
  • Available in a wide range of sizes
Specifications
Specifications
45mm
Gray
59.5 mm
Tapered Stopper
1/Ea.
50.5 mm
Natural Rubber
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Deutsch Neumann™ Grey Natural Rubber Stoppers > Bottom Diameter: 50.5mm; Height: 45mm