Learn More
Agilent BioTek Green Test Dye
Green Test Dye is used to test reagent injectors on all multi-mode readers during qualification and is for use with Microplate Washers and Readers.
Specifications
For Use With (Equipment) Agilent BioTek Microplate Washers and Readers
For Use With (Application) Test reagent injectors on all multi-mode readers during instrument qualification
Quantity 125 mL
View More Specs
Specifications
Agilent BioTek Microplate Washers and Readers
Test reagent injectors on all multi-mode readers during instrument qualification
125 mL
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Agilent BioTek Green Test Dye