missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP Bel-Art™ H-B Burac Plus Blood Bank Verification Thermometer; -5°C to 20°C
This H-B Burac Plus Blood Bank Verification Thermometer is mercury free, traceable to NIST and covers ranges from -5°C to 20°C (20°F to 70°F).
Brand: SP Bel-Art™ B60600-0700
This item is not returnable.
View return policy
Description
- Verify sample temperatures in Blood Bank refrigerators
- Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
- Insert thermometer into the bottle and place near the samples for best results
- Thermometers contain organic liquid, or EnviroKleen™ certified Enviro-Safe™ liquid; feature permanently fused markings and lead free glass
- PFA coating on select thermometers safely contain glass and liquid in case of breakage
- Manufactured and tested by our USA quality team; individually serialized
- Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
- Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available
Specifications
Glass Beads | |
4.9 in. | |
Organic Liquid | |
-5°C to 20°C |
30 mL | |
125 mm | |
Thermometer |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Sicherheitshinweis
Ergänzende Informationen
- H-B DURAC PLUS BLOOD BANK VERIFICATION THERMOMETER ; -5 TO 20C
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Sicherheitshinweis
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use CO2, powder, or water spray to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
Ergänzende Informationen
- MIXTURE LIST-Contains : acetone