Learn More
SP Bel-Art™ H-B Durac Plus Pocket Liquid-in-Glass Laboratory Thermometer; Original Liquid Fill
Featuring a case to protect from breakage, these H-B Durace Plus Pocket Liquid-in-Glass Laboratory Thermometers are a compact 160 mm (6.3 in.), making them ideal for field studies, waste water treatment and any 'on-the-go' applications.
18.60€ - 20.39€
Specifications
Immersion Type | Total Immersion |
---|---|
Length (Metric) | 160 mm |
Product Type | Thermometer |
Length (English) | 6.3 in. |
Product Code | Brand | Temperature Range (Metric) | Accuracy | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Accuracy | Description | Price | Quantity & Availability | |||
17511295
|
SP Bel-Art™
B60770-0400 |
-10°C to 100°C | ±1°C | Window Plastic Case |
18.60€
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
17521295
|
SP Bel-Art™
B60770-1100 |
-10°C to 100°C | ±1°C | Closed Plastic Case |
20.39€
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
17541295
|
SP Bel-Art™
B60770-1800 |
-10°C to 100°C | ±1°C | Closed Metal Case |
19.54€
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
Description
- Mercury free; contain blue organic liquid against lead-free, white back glass
- Tested and inspected by our USA quality team
- Traceable to NIST
- Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
- Case options include:
- Plastic Window - encased in plastic with an open window
- Closed Plastic - housed in removable plastic case
- Closed Metal - housed in a removable metal case
- Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Specifications
Total Immersion | |
160 mm | |
Thermometer | |
6.3 in. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- H-B DURAC PLUS POCKET LIQUID-IN-GLASS LAB THERMOME
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : acetone