Learn More
H-B Instrument™ Durac™ Plus™ Precision Verification Thermometers
To verify sample temperatures in freezers, refrigerators, incubators and ovens.
48.50€ - 107.00€
Specifications
Bottle Fill | Glass Beads |
---|---|
Certifications/Compliance | ISO/IEC 17025:2005, A2LA, NIST, DAkkS |
Includes | Multilanguage instructions, statement of accuracy indicating accuracy traceable to NIST |
Thermometer Fill | Blue Organic Liquid |
Product Type | Precision Verification Thermometer |
Includes: Multilanguage (English, French, German, Italian, Portuguese and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
Product Code | Brand | Bottle Size | Temperature Range (Metric) | For Use With (Application) | Length (Metric) Overall | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bottle Size | Temperature Range (Metric) | For Use With (Application) | Length (Metric) Overall | Price | Quantity & Availability | ||
17501285
|
H-B Instrument™
B60600-0600 |
60 mL | -5°C to +15°C | Refrigerator | 125 mm |
48.50€
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
17541285
|
H-B Instrument™
B60600-1000 |
30 mL | 15°C to 50°C | Incubator | 135 mm |
50.85€
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
17561275
|
H-B Instrument™
B60600-0200 |
60 mL | 15°C to 30°C | Incubator | 180 mm |
85.08€
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
17551275
|
H-B Instrument™
B60600-0100 |
60 mL | -2°C to +10°C | Refrigerator | 180 mm |
88.25€
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
17541275
|
H-B Instrument™
B60600-0000 |
60 mL | -25°C to +5°C | Freezers | 210 mm |
97.20€
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
17571275
|
H-B Instrument™
B60600-0300 |
60 mL | 95°C to 115°C | Oven | 200 mm |
76.67€
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
17581275
|
H-B Instrument™
B60600-0400 |
60 mL | 25°C to 45°C | Incubator | 190 mm |
107.00€
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||
Description
Blue liquid fill on white backed glass makes it easier to confirm temperature in freezers, refrigerators, incubators.
Applications:
Ideal for chemical industry, petrochemistry, pharmaceutical, medical, cosmetic, producers of chemicals, university, dairies, breweries, distillers, beer cellars, refrigeration house, hot houses, scientific institutes, blood banks, refrigeration/freezer/incubator/oven equipment producers and producers of
laboratory equipment.
Warranty
Five-year limited. See hbinstrument.com/conditions.php for details.
Specifications
Glass Beads | |
Multilanguage instructions, statement of accuracy indicating accuracy traceable to NIST | |
Precision Verification Thermometer |
ISO/IEC 17025:2005, A2LA, NIST, DAkkS | |
Blue Organic Liquid |
Safety and Handling
- H-B DURAC PLUS FREEZER VERIFICATION THERMOMETER; -
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use CO2, powder, or water spray to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : acetone