Learn More
R&D Systems™ Human 20S Immunoproteasome DuoSet ELISA
Human 20S Immunoproteasome DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand:  R&D Systems™ DY8194-05
Product Code. 18738717
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
The Human 20S Immunoproteasome DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human 20S Immunoproteasome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250.00 to 16000 pg/mL
Human
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Solid Phase Sandwich ELISA
1 Kit (5 Plates)
Product Title
R&D Systems™ Human 20S Immunoproteasome DuoSet ELISA > 1 Kit (for 5 Plates)