missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA Tag Monoclonal Antibody (2-2.2.14), HRP, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 26183HRP
This item is not returnable.
View return policy
Description
HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.Specifications
HA Tag | |
Monoclonal | |
1 mg/mL | |
proprietary buffer with 0.07% Kathon; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Tag | |
Antibody | |
IgG1 |
Western Blot | |
2-2.2.14 | |
HRP | |
HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin | |
HA peptide YPYDVPDYA derivitized to ovalbumin. | |
50 μL | |
Primary | |
4° C | |
Liquid | |
Tag |