Learn More
Gilac™ HACCP Food Contact Container
HACCP Food Contact Container is ideal for storage, food preparation, and transporting foodstuffs.
47.75€
Specifications
Product Type | HACCP Food Container |
---|---|
Material | Polyethylene |
Volume (English) | 13.2 gal. |
Volume (Metric) | 50 L |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
17948911
|
Gilac™
G613221 |
White |
47.75€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17988651
|
Gilac™
G613223 |
Yellow |
47.75€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17978511
|
Gilac™
G613228 |
Blue |
47.75€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17968911
|
Gilac™
G613233 |
Red |
47.75€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
17918701
|
Gilac™
G613236 |
Green |
47.75€
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
Description
- HACCP round container.
- Food contact certified.
- For storage, food preparation and transporting foodstuffs.
- Highly durable.
- Reinforced bottom with anti-wear skirt.
- Reinforced handles to prevent distortion during carrying.
- Optional, choice of two lids.
- External Diameter includes handles.
- Width corresponds to diameter without handles.
Specifications
HACCP Food Container | |
Polyethylene | |
13.2 gal. | |
50 L |