missing translation for 'onlineSavingsMsg'
Learn More

Hairy (Drosophila) Antibody (1/24) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

196.00€ - 382.00€

Specifications

Antigen Hairy (Drosophila)
Clone 1/24
Dilution Western Blot : 1:100 - 1:2000, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Applications Western Blot, Immunohistochemistry, ChIP Assay
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18664819
View Documents
Novus Biologicals
NBP2-76659-0.025ml
0.025 mL
196.00€
0.03mL
Please to purchase this item. Need a web account? Register with us today!
18083464
View Documents
Bio-Techne
NBP2-76659
0.1 mL
382.00€
0.10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hairy (Drosophila) Monoclonal specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
Specifications

Specifications

Hairy (Drosophila)
Western Blot : 1:100 - 1:2000, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Monoclonal
Purified
Cellular Signaling, Developmental Biology, Epigenetics, Stem Cells
PBS with 0.02% sodium azide
IgG1
Protein A purified
This antibody will stain Drosophila embryos.
1/24
Western Blot, Immunohistochemistry, ChIP Assay
Unconjugated
Mouse
Drosophila
Hairy gene product
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hairy (Drosophila) Antibody (1/24) - BSA Free, Novus Biologicals™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.