missing translation for 'onlineSavingsMsg'
Learn More

Haloperidol hydrochloride, Tocris Bioscience™

Product Code. 30001151
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Antagonist, partly D2 selective

Dopamine antagonist with selectivity for D2-like receptors (Ki values are 1.2, ∽ 7, 2.3, ∽ 80 and ∽ 100 nM for D2, D3, D4, D1 and D5 receptors respectively). Subtype-selective NMDA antagonist. Identified as targeting human host proteins that interact with SARS-CoV-2.

Specifications

CAS 1511-16-6
Molecular Formula C21H24Cl2FNO2
Quantity 100 mg
Synonym haloperidol hydrochloride, unii-um06w2adry, um06w2adry, haloperidol chloride, haloperidol chlorohydrate, haloperidol hydrochloride mi, c21h23clfno2.clh, regid_for_cid_11495267, 1-butanone, 4-4-4-chlorophenyl-4-hydroxy-1-piperidinyl-1-4-fluorophenyl-, hydrochloride 1:1
InChI Key JMRYYMBDXNZQMH-UHFFFAOYSA-N
SMILES C1CN(CCC1(C2=CC=C(C=C2)Cl)O)CCCC(=O)C3=CC=C(C=C3)F.Cl
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(4-fluorophenyl)butan-1-one;hydrochloride
Molecular Weight (g/mol) 412.326
PubChem CID 11495267
Formula Weight 412.33
Percent Purity >99%
Chemical Name or Material Haloperidol hydrochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'