missing translation for 'onlineSavingsMsg'
Learn More

CD148 Hamster anti-Mouse, PE, Clone: 8A-1, BD

Hamster Monoclonal Antibody

Brand:  BD Biosciences 565747

371.46 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15884499

  • 372.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Description

Description

The 8A-1 monoclonal antibody specifically binds to CD148, which is also known as Protein tyrosine phosphatase receptor type J (Ptprj), Receptor-type tyrosine-protein phosphatase eta (R-PTP-eta, PTP η ), or Density-enhanced phosphatase-1 (DEP-1). CD148 is a receptor-like tyrosine phosphatase that plays important roles in regulating cell growth, proliferation, differentiation, adhesion, and migration. It is expressed on a wide variety of cell types including epithelial cells, endothelial cells, and fibroblasts. For cells of hemopoietic origin, it is variably expressed on macrophages, neutrophils, dendritic cells, and on developing and mature B cells. CD148 is expressed on platelets and can regulate platelet activation. Although expressed at low levels on thymocytes and mature T cells, its expression is upregulated upon T cell receptor (TCR)-mediated stimulation and may play a role in regulating TCR signaling. CD148, along with CD45, are critical protein tyrosine phosphatases for regulating Src family kinase signaling networks in immune cells.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD148
Monoclonal
0.2mg/mL
Ptprj; R-PTP-J; BET; Byp; DEP-1; PTPbeta2; Ptpb2; R-PTP-eta; Scc-1; Scc1
Hamster
Affinity Purified
RUO
Mouse
IgG1
Flow Cytometry
8A-1
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse CD148 Transfected Cell Line
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD148 Hamster anti-Mouse, PE, Clone: 8A-1, BD > 50μg; PE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.