Learn More
Thermo Scientific Chemicals HANDY-LOK^t closure set for use with stock# 91717
Click to view available options
Quantity:
1 Ea.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- For use with 17 x 17 in. polyethylene glove bags
Specifications
Specifications
For Use With (Equipment) | Polyethylene Glove Bags (17 x 17 in.) |
Quantity | 1 Ea. |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Title
Thermo Scientific Chemicals HANDY-LOK^t closure set for use with stock# 91717 >