missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HARS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA557474
This item is not returnable.
View return policy
Description
Immunogen sequence: AVRCQSQVAE AVLTSQLKAH QEKPN Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 77%.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is an enzyme belonging to the class II family of aminoacyl-tRNA synthetases. Functioning in the synthesis of histidyl-transfer RNA, the enzyme plays an accessory role in the regulation of protein biosynthesis. The gene is located in a head-to-head orientation with HARS on chromosome five, where the homologous genes likely share a bidirectional promoter. Mutations in this gene are associated with the pathogenesis of Perrault syndrome, which involves ovarian dysgenesis and sensorineural hearing loss. Alternative splicing results in multiple transcript variants of this gene.Specifications
HARS2 | |
Polyclonal | |
Unconjugated | |
HARS2 | |
4631412B19Rik; D-tyrosyl-tRNA deacylase 1 homolog; EC 6.1.1.21; EGK_16929; HARS2; Hars2l; HARSL; HARSR; HARS-related; HisRS; histidine translase; histidine tRNA ligase 2, mitochondrial (putative); histidine-tRNA ligase homolog; Histidine--tRNA ligase-like; Histidyl-tRNA synthetase; histidyl-tRNA synthetase 2; histidyl-tRNA synthetase 2, mitochondrial; histidyl-tRNA synthetase 2, mitochondrial (putative); histidyl-tRNA synthetase 2, mitochondrial, hisRS; histidyl-tRNA synthetase-like; HO3; PRLTS2; Probable histidine--tRNA ligase, mitochondrial; probable histidyl-tRNA synthetase, mitochondrial; probable histidyl-tRNA synthetase, mitochondrial-like; putative histidyl-tRNA synthetase, mitochondrial; RGD1308426; zinc finger, matrin type 2; Zmat2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23438 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49590 | |
HARS2 | |
Recombinant protein corresponding to Human HARS2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction