missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Hazardous Substances Rack
The Hazardous Substances Rack provides storage of aggressive chemicals.
Brand: asecos™ 27857
This item is not returnable.
View return policy
Description
- Galvanised rack frames
- Sturdy construction due to reinforcement cross bars
- Storage levels in 25 mm increments, height-adjustable
- Load capacity per storage shelf: 200 kg (evenly distributed)
- Rack system can be extended individually
- Flat-pack delivery with assembly instructions
- Varioius storage level options
- Polyethylene sumps with high resistance against aggressive chemicals (such as acids and alkalis)
Specifications
7.9 gal. | |
Steel | |
41.73 in. | |
41.73 in. | |
78.74 in. | |
25.08 in. | |
156.53 lb. |
30 L | |
Drum Rack | |
1060.00 mm | |
1060.00 mm | |
2000.00 mm | |
637.00 mm | |
71 kg |