Learn More
IKA™ HBC 10 Basic Heating Bath
Brand: IKA™ 0004135000
3724.56 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
20°C to 200°C |
Product Content Correction
Product Title
IKA™ HBC 10 Basic Heating Bath > Temperature Range: 20 to 200°C