Learn More
Horst™ HBST Model Heating Tape
Brand:  Horst™ 20701
 View more versions of this product
Product Code. 10057071
  • 133.00€ / Each
Estimated Shipment: 05-05-2025
Specifications
1 Lm
1 m
HBST
1 Lm
∼230V Voltage
Heating Tape
Certificates
View accessory products
Product Title
Horst™ HBST Model Heating Tape > Length: 1m