Learn More
behr Labor-Technik™ HCL Absorber
Expel chloride when determining the COD of sea water. behr Labor-Technik™ HCL Absorbers are ideal for COD testing at more than 300mg in the COD range of 5 to 50mg
Brand: behr Labor-Technik™ B00217890
This item is not returnable.
View return policy
Description
- For expelling chlorine with chlorine content above 1000 mg and COD in the range above 50mg
- No plug
Specifications
HCL Absorber | |
Determining The COD of Sea Water | |
HCl 29 |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
behr Labor-Technik™ HCL Absorber > For Use With (Application): Determining the COD of Sea Water