Learn More
behr Labor-Technik™ HCL Absorber
Expel chloride when determining the COD of sea water. behr Labor-Technik™ HCL Absorbers are ideal for COD testing at more than 300mg in the COD range of 5 to 50mg
Brand:  behr Labor-Technik™ B00217890
Product Code. 10517332
  • 138.00€ / Each
Estimated Shipment: 16-05-2025
Description
  • For expelling chlorine with chlorine content above 1000 mg and COD in the range above 50mg
  • No plug
Prior to insertion into the reaction vessel, either fill the filter cartridge with absorbent material (calcium hydroxide) or soak it with distilled water. The solution is either stirred for several hours on the standalone magnetic agitator or on the magnetic series agitator with 12 positions, then to be driven off as Cl.
Specifications
Specifications
HCL Absorber
Determining The COD of Sea Water
HCl 29
Product Title
behr Labor-Technik™ HCL Absorber > For Use With (Application): Determining the COD of Sea Water