Learn More
Tocris Bioscience™ HDAC4 CHDI Degrader 11
Potent and selective HDAC4 Degrader (PROTAC™)
Brand:  Tocris Bioscience™ 7882/5
Product Code. 18044052
  • 335.00€ / 5mg
Estimated Shipment: 30-01-2025
Description
HDAC4 CHDI Degrader 11 is a potent and selective HDAC4 Degrader (PROTAC™) (DC50 values are 4 and 6 nM in Jurkat E6-1 and Jurkat cells respectively). Comprises the class IIa HDAC inhibitor trifluoromethyloxadiazole joined by a linker to a ligand for Von Hippel-Lindau (VHL) protein. In a mouse cell model of Huntingtons disease, HDAC4 CHDI Degrader 11 potently degrades HDAC4 (DC50 = 1nM). Can be used with P-glycoprotein inhibitor Elacridar (Cat. No. 4646) for more effective degradation in neuroblastoma cell lines. HDAC4 antibody validated for Simple Western™ (automated Western) instruments also available: Catalog # NBP2-22151 .PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
(2S,4R)-1-((3R,26S)-26-(tert-Butyl)-3-methyl-1,24-dioxo-5-propyl-1-(4-(5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl)phenyl)-10,13,16,19,22-pentaoxa-2,5,25-triazaheptacosan-27-oyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide
Active Degraders
98%
5 mg
C52H73F3N8O11S