Learn More
Verenigde Kunststof Bedrijven™ HDPE Barrel
Verenigde Kunststof Bedrijven™ HDPE Barrel is designed for storage, handling and transportation of products manufactured by the chemical, petrochemical and food industries.
38.45€
Specifications
Product Type | Barrel |
---|---|
Certifications/Compliance | UN Approved |
Closure Type | Removable Lid |
Color | Blue |
Material | HDPE |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10257354
|
Verenigde Kunststof Bedrijven™
602900 |
30 L | |||||||
Description
- HDPE UN approved Lid barrels
- Removable black Lid
- Metal fastener
- For transport and storage Chemical, Petrochemical, Food Industries
- Complete Draining
- enabling re-use or recycling
- Does not age or corrode
- Easy to Stack and pallatise
Specifications
Barrel | |
Removable Lid | |
HDPE |
UN Approved | |
Blue |