Learn More
3M™ Heavy Duty EPDM Breathing Tube
Extended fixed-length breathing tube made from EPDM rubber.
Brand:  3M™ 7100265244
 View more versions of this product
Product Code. 17796993
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • Suitable for use with specific 3M™ Versaflo™ headtops including M-200, M-300 and S Series Hoods.
  • EPDM rubber hose
  • 100 cm length
  • Protective breathing tube covers available to lengthen service life of breathing tubes