Learn More
3M™ Heavy Duty EPDM Breathing Tube, SS-BT-44
3M™ Heavy Duty EPDM Breathing Tube, SS-BT-44 is a fixed-length EPDM rubber breathing tube designed for use with certain approved 3M™ Powered Air Systems.
Brand:  3M™ 7100236824
Product Code. 17746973
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • Fixed-length breathing tube made from EPDM rubber
  • Approved for use with 3M™ Powered Air Respirator PF-600E
  • Suitable for use with specific 3M™ Versaflo™ headtops when using the 3M™ Adapter Assembly QRS Thread ADP-03.
  • 80 cm length
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EPDM Rubber