Learn More
Brady™ Heavy Duty Nylon Lockout Belt Pouch
Simply fill the pouch with the lockout devices you need and have them handy when they are required. The Heavy Duty Nylon Lockout Belt Pouch is supplied with loops for hanging on a belt.
Brand:  Brady™ 051172
Product Code. 11791865
  • 78.05€ / Each
Estimated Shipment: 16-06-2025
Description
  • Contents not included.
  • Dimensions: 50.8 x 177.8 x 127 mm (D x W x H)
Specifications
Specifications
Lockout Belt Pouch
LOCKOUT BELT POUCH, RED 5 X7 in.
Black on Red
1 Ea.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Brady™ Heavy Duty Nylon Lockout Belt Pouch > Lockout Belt Pouch; Dimensions (D x W x H): 50 x 190 x 120 mm