Learn More
Heidolph™ Instruments Hei-VAP Ultimate HL G6B, EU-plug
The Hei-VAP Ultimate HL G6B allows for precise setting of rotation speed and heating bath temperature on the digital touch display or directly via the knobs on the operating panel.
Brand:  Heidolph™ Instruments 575-01610-00
Product Code. 17313350
  • 6485.00€ / Each
Estimated Shipment: 08-07-2025
Add to basket
Description
Description
Precise setting of rotation speed and heating bath temperature on the digital touch display or – for quick access during an active process – directly via the knobs on the operating panel.
  • Central operating panel with 7 in. touch display
  • Hand lift model with transparent plastic coated G6 vertical glassware for reflux distillation
  • Clear display of all values (Basic and Advanced modes)
  • With timer, limitation of max. temperature, multilingual menu and text display for error messages
  • Separate knobs for rotation and heating with locking function to prevent accidental readjusting
  • LED ring lights and pictogram to display the activity even from a distance
  • Integrated solvent library to facilitate adjustment of the required vacuum on an external vacuum controller
  • USB and Micro SD plus LAN / RS 232 interfaces (Lab 4.0 ready)
  • Optional display of vapor temperature
  • Subsequent upgrade to Hei-VAP Ultimate Control possible, including vacuum and chiller control
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 L
Hand Lift Rotary Evaporator
EU-plug
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Heidolph™ Instruments Hei-VAP Ultimate HL G6B, EU-plug > Type: Hand Lift Rotary Evaporator; Capacity (Metric): 5 L