Learn More
CleanSpace™ Technology Helmet Hook Strap Accessory Product Code.: 15960072
Estimated Shipment: 13-08-2025
Only null left
Add to basket
CleanSpace™ Technology Helmet Hook Strap Accessory
Product Code. 15960072 Shop All CleanSpace Technology Products
Dimensions (L x W x H): 950 x 350 x 600 mm
Click to view available options
:
Dimensions (L x W x H): 950 x 350 x 600 mm
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15960072
Brand: CleanSpace™ Technology PAF0075
Only null left
Estimated Shipment: 13-08-2025
Add to basket
Only null left
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Helmet hook strap accessory is designed for use when wearing the respirator with a safety helmet.
  • Used with the revolutionary CleanSpace™ Respirator: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
Specifications
Disposable Reusable
Product Type Helmet Hook Strap
For Use With (Application) Welding, Woodworking, Manufacturing, Smelting, Construction, Recycling Plants, Emergency Services, Mining, Agriculture, Processing Plants, Grinding
For Use With (Equipment) PAF-1014, PAF-1106
Model PAF-0075
Storage Requirements Store at -10°C to +55°C
Dimensions (L x W x H) 950 x 350 x 600 mm
Weight (Metric) 25 g
Product Content Correction
Product Title
CleanSpace™ Technology Helmet Hook Strap Accessory >