Learn More
Brady™ High Adhesion General Identification Polyester Labels
Rough, glossy and irregular surfaces are no match for the ToughBond™ ultra aggressive polyester label material (B-483). Print almost anywhere with a compact Brady™ handheld label maker.
32.20€ - 84.50€
Specifications
Adhesive Type Permanent Rubber
Color Black on White
Finish Gloss
Format Die-cut
Material Polyester
View More Specs
Products 3
Product Code Brand Dimensions (LxW) Price Quantity & Availability  
Product Code Brand Dimensions (LxW) Price Quantity & Availability  
17930730
View Documents Promotion Details
Brady™
170773
0.5 x 1 in.
80.10€
Pack of 480
In Stock
to see stock.
Add to basket
17910730
View Documents
Brady™
170771
2 x 0.5 in.
32.20€
Pack of 80
In Stock
to see stock.
Add to basket
17940730
View Documents Promotion Details
Brady™
170772
2 x 1 in.
84.50€
Pack of 140
In Stock
to see stock.
Add to basket
Description
Description
  • Ideal for general identification and marking powder-coated and smooth plastic surfaces
  • Resistant to abrasions, extreme temperatures and solvents such as fuel, oil and water
  • Made with a permanent rubber-based adhesive that secures tightly to flat textured metals and smooth plastics alike
Specifications
Specifications
Permanent Rubber
Gloss
Polyester
Non-sterile
BMP41, BMP51, M511
0.004 in.
Black on White
Die-cut
Labels
Asset Tracking, Barcode Labeling, Component and Equipment Labeling, Data and Telecommunications Labeling, Electrical Labeling, General Identification, Inventory and Inspection Labeling
-40°F to +248°F