Learn More
Greiner Bio-One High Grade Polypropylene Pipet Tips
Manufactured from high-grade polypropylene to give the optimum performance and fit. Greainer BioOne™ High Grade Polypropylene Pipet Tips are ideally suited for Gilson™ Pipetman models.
Brand:  Greiner Bio-One 745290
 View more versions of this product
Product Code. 11789542
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
  • High breaking strength, dimensionally stable, heat-resistant up to approx. +140°C; cold-resistant down to -190°C
  • Autoclavable
  • Free of heavy metals
  • Conforms to the ISO 9001 standard
  • Tested for conformance with the international standard DIN ISO 8655
  • Liquid end is bevelled so less liquid remains on pipet tip, contact surface of pipetted liquids is reduced and pipetting accuracy increases
  • Ideally suited for Gilson™ Pipetman P20, P100, P200, F5, F10, F25, F50, F200, U200
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 to 5000 μL
Autoclavable
Bulk
Polypropylene
Natural
ISO 9001, DIN ISO 8655
Socorex™ Pipette, Gilson™ Pipetman™ P5000