Learn More
Idex High-Pressure, Zero Dead Volume Unions
PEEK ZDV unions provide accurate results
63.28€ - 70.96€
Specifications
Type Union
Pressure Rating 6000 psi
Material PEEK
View More Specs
Products 2
Product Code Brand Diameter (English) Inner Swept Volume Price Quantity & Availability  
Product Code Brand Diameter (English) Inner Swept Volume Price Quantity & Availability  
13010410
View Documents
Idex
02014-37
0.01 in. 0.071 ÎĽL
63.28€
Each
Estimated Shipment: 04-07-2025
Add to basket
13040390
View Documents
Idex
02014-29
0.02 in. 0.283 ÎĽL
70.96€
Each
Estimated Shipment: 04-08-2025
Add to basket
Description
Description
  • Inert PEEK fluid path
  • 10-32 coned configuration
Specifications
Specifications
Union
6000 psi
PEEK
Documents
Product Certifications
Product Content Correction
Product Title
Idex High-Pressure, Zero Dead Volume Unions