missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Histone Acetyltransferase Inhibitor IV, CPTH2

Product Code. 15427139
Click to view available options
Quantity:
5 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Histone acetylation plays an important role in regulating the chromatin structure and is tightly regulated by two classes of enzyme, histone acetyltransferases (HAT) and histone deacetylases (HDAC). Deregulated HAT and HDAC activity plays a role in the development of a range of cancers. Consequently, inhibitors of these enzymes have potential as anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357649-93-5
Molecular Formula C14H14ClN3S
Synonym HAT Inhibitor IV; Gcn5p Inhibitor

Specifications

Quantity 5 mg
Description Brown
Content And Storage 2°C to 8°C
For Use With (Application) A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators
Formula Weight 291.80g/mol
Sensitivity Light Sensitive
Solubility Soluble in DMSO
Product Type Signal Transduction Reagent
Form Solid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.