missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Specifications
Antigen | Histone H3 |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18392334
|
Bio-Techne
NBP3-03269-20UL |
20 μL |
198.00€
20µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
18312457
|
Bio-Techne
NBP3-03269-100UL |
100 μL |
468.00€
100µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Histone H3 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-136 of human Histone H3 (NP_002098.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
PBS with 50% glycerol, pH7.3. | |
126961 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title